Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A 83-01 sodium | 2828431-89-4 | 98.4% | 444.51 g/mol | C25H19N5NaS | 100 MG
SDP

Catalog No. 5000241742
Encompass_Preferred

A 83-01 sodium is a sodium salt small-molecule inhibitor of the TGF-β type I receptor family (ALK4, ALK5, ALK7) intended for biochemical and cellular research. It is supplied as a high-purity solid with low-nanomolar potency and is typically handled as a DMSO-soluble stock and stored protected from moisture and light.

  • Potent ALK4/5/7 inhibition with IC50s ≈12 nM (ALK5), 45 nM (ALK4), 7.5 nM (ALK7).
  • High purity suitable for research use.
  • Sodium salt form for improved handling and solubility in polar solvents.
  • Solid, white to light yellow physical form for easy weighing and storage.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes, including 100 mg.

Catalog No. 50-002-41742 Supplier Medchemexpress LLC Supplier No. HY10432A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 83-01 sodium is a sodium salt small-molecule inhibitor of the TGF-β type I receptor family (ALK4, ALK5, ALK7) intended for biochemical and cellular research. It is supplied as a high-purity solid with low-nanomolar potency and is typically handled as a DMSO-soluble stock and stored protected from moisture and light.

  • Potent ALK4/5/7 inhibition with IC50s ≈12 nM (ALK5), 45 nM (ALK4), 7.5 nM (ALK7).
  • High purity suitable for research use.
  • Sodium salt form for improved handling and solubility in polar solvents.
  • Solid, white to light yellow physical form for easy weighing and storage.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes, including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.