Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A 83-01 sodium | 2828431-89-4 | MFCD08705403 | 98.4% | 444.51 | C25H19N5NaS | 1 MG
SDP

Catalog No. 5000243467
Encompass_Preferred

A 83-01 sodium is the sodium salt form of a potent TGF-β type I receptor inhibitor used in biochemical and cell-based research to block ALK5-, ALK4-, and ALK7-mediated signaling. The compound is provided as a solid research reagent with high solubility in DMSO and guidance for in vivo formulation and storage. It is intended for research use only.

  • Potent inhibitor of TGF-β type I receptors ALK5, ALK4, and ALK7.
  • Sodium salt form improves aqueous formulation options.
  • High purity suitable for research applications (98.4%).
  • Solubility in DMSO ≥100 mg/mL for in vitro use.
  • Includes in vivo formulation guidance for suspension dosing.
  • Store sealed at 4°C away from moisture and light.
  • Molecular formula C25H19N5NaS and molecular weight 444.51 g/mol.

Catalog No. 50-002-43467 Supplier Medchemexpress LLC Supplier No. HY10432A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 83-01 sodium is the sodium salt form of a potent TGF-β type I receptor inhibitor used in biochemical and cell-based research to block ALK5-, ALK4-, and ALK7-mediated signaling. The compound is provided as a solid research reagent with high solubility in DMSO and guidance for in vivo formulation and storage. It is intended for research use only.

  • Potent inhibitor of TGF-β type I receptors ALK5, ALK4, and ALK7.
  • Sodium salt form improves aqueous formulation options.
  • High purity suitable for research applications (98.4%).
  • Solubility in DMSO ≥100 mg/mL for in vitro use.
  • Includes in vivo formulation guidance for suspension dosing.
  • Store sealed at 4°C away from moisture and light.
  • Molecular formula C25H19N5NaS and molecular weight 444.51 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.