Learn More
Medchemexpress LLC A 83-01 sodium 5mg | 2828431-89-4 | 5MG
SDP
Catalog No. 5000243092
Encompass_Preferred
A 83-01 sodium is a potent inhibitor of TGF- type I receptor ALK5 kinase ALK4 and ALK7 with IC50s of 12 nM 45 nM and 7 5 nM against the transcription induced by ALK5 ALK4 and ALK7 respectively[1
Catalog No. 50-002-43092 Supplier Medchemexpress LLC Supplier No. HY10432A5MG
A 83-01 sodium is a potent inhibitor of TGF- type I receptor ALK5 kinase ALK4 and ALK7 with IC50s of 12 nM 45 nM and 7 5 nM against the transcription induced by ALK5 ALK4 and ALK7 respectively[1