Learn More
Medchemexpress LLC A-836339 | 959746-77-1 | 99.8% | 1 ML
SDP
Catalog No. 5000342199
Encompass_Preferred
A-836339 is a selective CB2 receptor agonist with Ki values of 0.4 nM in humans and 0.8 nM in rats. It demonstrates various effects, including analgesia, gastric protection, anti-inflammation, and antioxidant activity. This compound achieves antinociceptive and analgesic effects by activating CB2 receptors located in the dorsal root ganglia and spinal cord.
  • Provides gastric protective benefits through anti-inflammatory and antioxidant mechanisms.
  • Can be used as a CB2-specific radioligand for autoradiography and PET imaging.
  • Suitable for research on inflammatory pain, neuropathic pain, gastric ulcers, and cerebral ischemia.
Catalog No. 50-003-42199 Supplier Medchemexpress LLC Supplier No. HY1276110MM/1ML
Add to Cart
Add to Cart
A-836339 is a selective CB2 receptor agonist with Ki values of 0.4 nM in humans and 0.8 nM in rats. It demonstrates various effects, including analgesia, gastric protection, anti-inflammation, and antioxidant activity. This compound achieves antinociceptive and analgesic effects by activating CB2 receptors located in the dorsal root ganglia and spinal cord.
  • Provides gastric protective benefits through anti-inflammatory and antioxidant mechanisms.
  • Can be used as a CB2-specific radioligand for autoradiography and PET imaging.
  • Suitable for research on inflammatory pain, neuropathic pain, gastric ulcers, and cerebral ischemia.