Learn More
Miele, Inc. A 845 injector nozzle Ø 2.5 x 125 mm (1 off)
Catalog No. 501713081
Encompass_Preferred
A 845 Injector nozzle
With plastic supports, nozzle diameter 2.5, length 125 mm, 1 item.
Suitable for mobile injector units & baskets, w/ screw connection
Part of the SimpleLoad system
Packaging unit with 1 nozzle
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3081 Supplier Miele, Inc. Supplier No. 11056670
Add to Cart
Add to Cart
A 845 Injector nozzle
With plastic supports, nozzle diameter 2.5, length 125 mm, 1 item.
Suitable for mobile injector units & baskets, w/ screw connection
Part of the SimpleLoad system
Packaging unit with 1 nozzle
Visit www.mieleusa.com/professional for technical information.