Learn More
Miele, Inc. A 846 Injector nozzle for bott
Catalog No. 502870753
Encompass_Preferred
A 846 Injector nozzle for bottles up to 10L and round flasks up to 6L (for use on A313 or A612)
Catalog No. 50-287-0753 Supplier Miele, Inc. Supplier No. 12665100
A 846 Injector nozzle for bottles up to 10L and round flasks up to 6L (for use on A313 or A612)