Learn More
Miele, Inc. A 846
Catalog No. 501953767
Encompass_Preferred
A 846 InjectorInjector nozzle with support, particularly suitable for laboratory bottles and round flasks.Long nozzle, 127 mm, total length with support 158 mm, Ø 10 mm
Catalog No. 50-195-3767 Supplier Miele, Inc. Supplier No. 11025590
A 846 InjectorInjector nozzle with support, particularly suitable for laboratory bottles and round flasks.Long nozzle, 127 mm, total length with support 158 mm, Ø 10 mm