Learn More
Miele, Inc. A 847
Catalog No. 501953768
Encompass_Preferred
A 847 InjectorInjector nozzle with support, particularly suitable for Erlenmeyer flasks.Nozzle length 127 mm, Ø 10 mm and support Ø mm, diameter of hexagonal support 142 mm
Catalog No. 50-195-3768 Supplier Miele, Inc. Supplier No. 12665110
A 847 InjectorInjector nozzle with support, particularly suitable for Erlenmeyer flasks.Nozzle length 127 mm, Ø 10 mm and support Ø mm, diameter of hexagonal support 142 mm