Learn More
Miele, Inc. A 848
Catalog No. 501953769
Encompass_Preferred
A 848 InjectorInjector nozzle with support, particularly suitable for measuring flasks.Nozzle length 350 mm, Ø 10 mm and support Ø mm, diameter of hexagonal support 142 mm
Catalog No. 50-195-3769 Supplier Miele, Inc. Supplier No. 11025610
A 848 InjectorInjector nozzle with support, particularly suitable for measuring flasks.Nozzle length 350 mm, Ø 10 mm and support Ø mm, diameter of hexagonal support 142 mm