Learn More
Medchemexpress LLC A 85380 hydrochloride | 174740-86-4 | 97.0% | 237.13 g/mol | C9H14Cl2N2O | 5 MG
SDP
Catalog No. 5000263774
Encompass_Preferred
A 85380 hydrochloride is a selective neuronal nicotinic acetylcholine receptor agonist with preference for the α4β2 subtype. Supplied as the hydrochloride salt in small research quantities, it is intended for in vitro pharmacology and receptor profiling studies and formulation work.
Catalog No. 50-002-63774 Supplier Medchemexpress LLC Supplier No. HY1101315MG
A 85380 hydrochloride is a selective neuronal nicotinic acetylcholine receptor agonist with preference for the α4β2 subtype. Supplied as the hydrochloride salt in small research quantities, it is intended for in vitro pharmacology and receptor profiling studies and formulation work.