Learn More
Miele, Inc. A 857
Catalog No. 501953801
Encompass_Preferred
A 857 Conversation KitValue preservation through continued use of existing load carriersMagnet integrated into skidFor all laboratory load carriers on models G 7827, G 7828, PG 8527 and PG 8528.H 27, W 25, D 390 mm
Catalog No. 50-195-3801 Supplier Miele, Inc. Supplier No. 11235960
A 857 Conversation KitValue preservation through continued use of existing load carriersMagnet integrated into skidFor all laboratory load carriers on models G 7827, G 7828, PG 8527 and PG 8528.H 27, W 25, D 390 mm