Learn More
Miele, Inc. A 860 holder rack 2x4
Catalog No. 501713058
Encompass_Preferred
A 860 Holding frames
For injector modules A 300/2 and A 300/3
Arrangement: 2 x 4 fields
For use on A 300/2 and A 300/3 injector modules
For the optimum positioning of laboratory glassware
Part of the SimpleLoad system
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3058 Supplier Miele, Inc. Supplier No. 11056800
Add to Cart
Add to Cart
A 860 Holding frames
For injector modules A 300/2 and A 300/3
Arrangement: 2 x 4 fields
For use on A 300/2 and A 300/3 injector modules
For the optimum positioning of laboratory glassware
Part of the SimpleLoad system
Visit www.mieleusa.com/professional for technical information.