Learn More
Miele, Inc. A 860 Holding frame 2x4 suitab
Catalog No. 502870790
Encompass_Preferred
A 860 Holding frame 2x4 suitable for A300/3, A300/2
Catalog No. 50-287-0790 Supplier Miele, Inc. Supplier No. 12665410
A 860 Holding frame 2x4 suitable for A300/3, A300/2