Learn More
Miele, Inc. A 862 holder rack 4x8
Catalog No. 501713060
Encompass_Preferred
A 862 Holding frames
For injector modules A 302/2 and A 302/3
Arrangement: 4 x 8 fields
For use on A 302/2 and A 302/3 injector modules
For the optimum positioning of laboratory glassware
Plastic-coated
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3060 Supplier Miele, Inc. Supplier No. 11056820
A 862 Holding frames
For injector modules A 302/2 and A 302/3
Arrangement: 4 x 8 fields
For use on A 302/2 and A 302/3 injector modules
For the optimum positioning of laboratory glassware
Plastic-coated
Visit www.mieleusa.com/professional for technical information.