Learn More
Medchemexpress LLC HY-12149 10mg Medchemexpress, A-867744 CAS:1000279-69-5 Purity:>98%
SDP
Catalog No. 502080968
Encompass_Preferred
Medchemexpress, HY-12149 10mg A-867744 CAS:1000279-69-5 A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0968 Supplier Medchemexpress LLC Supplier No. HY1214910MG
Medchemexpress, HY-12149 10mg A-867744 CAS:1000279-69-5 A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.