Learn More
Medchemexpress LLC HY-100080 5mg , A-887826 CAS:1266212-81-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000805MG
Encompass_Preferred
HY-100080 5mg A-887826 CAS: 1266212-81-0 A-887826 is a potent, selective, oral bioavailable and voltage-dependent Na(v)1.8 sodium channel blocker with an IC50 of 11 nM . A-887826 attenuates neuropathic tactile allodynia in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1605