Learn More
Medchemexpress LLC A-908292 | 903886-95-3 | 99.9% | 360.43 | 1 ML
SDP
Catalog No. 5000362119
Encompass_Preferred
A-908292 is a potent and selective acetyl-CoA carboxylase 2 (ACC2) inhibitor with an IC50 of 23 nM for human ACC2. It is used in research on fatty acid metabolism and is also a click chemistry reagent.
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reduces serum glucose and triglyceride levels in ob/ob mice
  • Markedly reduces plasma triglyceride levels in ACC2 knockout mice
  • Stimulates the PPAR-α-dependent signaling pathway in rats
Catalog No. 50-003-62119 Supplier Medchemexpress LLC Supplier No. HY14700410MM/1ML
A-908292 is a potent and selective acetyl-CoA carboxylase 2 (ACC2) inhibitor with an IC50 of 23 nM for human ACC2. It is used in research on fatty acid metabolism and is also a click chemistry reagent.
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reduces serum glucose and triglyceride levels in ob/ob mice
  • Markedly reduces plasma triglyceride levels in ACC2 knockout mice
  • Stimulates the PPAR-α-dependent signaling pathway in rats