Learn More
Medchemexpress LLC HY-10038 5mg Medchemexpress, A 922500 CAS:959122-11-3 Purity:>98%
SDP
Catalog No. 501967726
Encompass_Preferred
Medchemexpress, HY-10038 5mg A 922500 CAS:959122-11-3 A 922500 (DGAT-1 Inhibitor 4a) is a potent, selective, and orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor with IC50s of 9 and 22 nM against human and mouse DGAT-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7726 Supplier Medchemexpress LLC Supplier No. HY100385MG
Medchemexpress, HY-10038 5mg A 922500 CAS:959122-11-3 A 922500 (DGAT-1 Inhibitor 4a) is a potent, selective, and orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor with IC50s of 9 and 22 nM against human and mouse DGAT-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.