Learn More
Medchemexpress LLC A 922500 | 959122-11-3 | 99.3% | 428.48 g·mol⁻¹ | C26H24N2O4 | 1 MG
SDP
Catalog No. 5000214215
Encompass_Preferred
A 922500 is a potent, selective, orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor used as a research reagent for biochemical and in vivo studies of lipid metabolism and pharmacology. It exhibits low-nanomolar activity against human and mouse DGAT-1 and is provided with validated purity and characterization data.
  • Potent, low-nanomolar DGAT-1 inhibition (IC50 ≈ 9 nM human, 22 nM mouse).
  • High purity suitable for research applications.
  • Available as powder and ready-to-use DMSO solution formats.
  • Characterized with COA, LC-MS, HNMR, and RP-HPLC data.
  • Soluble in DMSO and formulated for in vivo use per published protocols.
  • Stable under recommended frozen and refrigerated storage conditions.
Catalog No. 50-002-14215 Supplier Medchemexpress LLC Supplier No. HY100381MG
A 922500 is a potent, selective, orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor used as a research reagent for biochemical and in vivo studies of lipid metabolism and pharmacology. It exhibits low-nanomolar activity against human and mouse DGAT-1 and is provided with validated purity and characterization data.
  • Potent, low-nanomolar DGAT-1 inhibition (IC50 ≈ 9 nM human, 22 nM mouse).
  • High purity suitable for research applications.
  • Available as powder and ready-to-use DMSO solution formats.
  • Characterized with COA, LC-MS, HNMR, and RP-HPLC data.
  • Soluble in DMSO and formulated for in vivo use per published protocols.
  • Stable under recommended frozen and refrigerated storage conditions.