Learn More
TARGETMOL CHEMICALS INC A-935142 5MG
SDP
Catalog No. 502581312
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A-935142 is a human ether-a-go-go-related gene (hERG Kv 11.1) channel activator. a-935142 enhances hERG currents by slowing inactivation promoting activation reducing inactivation shortening atrial and ventricular repolarization and in a complex manner. purity: 100%
Catalog No. 50-258-1312 Supplier TARGETMOL CHEMICALS INC Supplier No. T140775MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A-935142 is a human ether-a-go-go-related gene (hERG Kv 11.1) channel activator. a-935142 enhances hERG currents by slowing inactivation promoting activation reducing inactivation shortening atrial and ventricular repolarization and in a complex manner. purity: 100%