Learn More
Abcam A 943931 dihydrochloride, H4 receptor antagonist, 10MG
Catalog No. 502583309
Encompass_Preferred
MW 368.3 Da, Purity >99%. Potent and selective H4 receptor antagonist (pKi = 7-8). Shows anti-inflammatory and analgesic effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3309 Supplier Abcam Supplier No. AB14280910MG
MW 368.3 Da, Purity >99%. Potent and selective H4 receptor antagonist (pKi = 7-8). Shows anti-inflammatory and analgesic effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement