Learn More
Medchemexpress LLC A-966492 10mM 1mL | 934162-61-5 | 10MM 1ML
SDP
Catalog No. 5000242723
Encompass_Preferred
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1 5 nM respectively
Catalog No. 50-002-42723 Supplier Medchemexpress LLC Supplier No. HY1061410MM 1ML
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1 5 nM respectively