Learn More
Selleck Chemical LLC A-966492 S2197-10mM/1mL
SDP
Catalog No. 503111767
Encompass_Preferred
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1 5 nM respectively
Catalog No. 50-311-1767 Supplier Selleck Chemical LLC Supplier No. S219710MM/1ML
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1 5 nM respectively