Learn More
Selleck Chemical LLC A-966492 5mg 934162-61-5
SDP
Supplier:  Selleck Chemical LLC S21975MG
Encompass_Preferred
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8037