Learn More
Medchemexpress LLC HY-10614 5mg Medchemexpress, A-966492 CAS:934162-61-5 Purity:>98%
SDP
Catalog No. 501968321
Encompass_Preferred
Medchemexpress, HY-10614 5mg A-966492 CAS:934162-61-5 A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8321 Supplier Medchemexpress LLC Supplier No. HY106145MG
Medchemexpress, HY-10614 5mg A-966492 CAS:934162-61-5 A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.