Learn More
Enzo Life Sciences A-967079 (1g). CAS: 1170613-55-4
SDP
Catalog No. 502013534
Encompass_Preferred
Potent and selective TRPA1 channel blocker. Exhibits greater than 1000-fold selectivity for TRPA1 over other TRP channels. Exhibits analgesic actions. Alternative name: (1E,3E)-1-(4-Fluorophenyl)-2-methyl-1-penten-3-one oxime. Purity: ≥99% (TLC). Solubility: Soluble in DMSO, DMF or ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-3534 Supplier Enzo Life Sciences Supplier No. ENZCHM2101000
Potent and selective TRPA1 channel blocker. Exhibits greater than 1000-fold selectivity for TRPA1 over other TRP channels. Exhibits analgesic actions. Alternative name: (1E,3E)-1-(4-Fluorophenyl)-2-methyl-1-penten-3-one oxime. Purity: ≥99% (TLC). Solubility: Soluble in DMSO, DMF or ethanol. Long Term Storage: -20°C.