Learn More
Medchemexpress LLC HY-108463 5mg Medchemexpress, A-967079 CAS:1170613-55-4 Purity:>98%
SDP
Catalog No. 501968552
Encompass_Preferred
Medchemexpress, HY-108463 5mg A-967079 CAS:1170613-55-4 A-967079 is a selective TRPA1 receptor antagonist with IC50s of 67 nM and 289 nM at human and rat TRPA1 receptors, respectively, and has good penetration into the CNS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8552 Supplier Medchemexpress LLC Supplier No. HY1084635MG
Medchemexpress, HY-108463 5mg A-967079 CAS:1170613-55-4 A-967079 is a selective TRPA1 receptor antagonist with IC50s of 67 nM and 289 nM at human and rat TRPA1 receptors, respectively, and has good penetration into the CNS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.