Learn More
Medchemexpress LLC A aggregation-IN-1 250mg
SDP
Catalog No. 5000206021
Encompass_Preferred
A aggregation-IN-1 (Compound 1b) is an amyloid-beta precursor protein inhibitor A aggregation-IN-1 inhibits amyloid-beta aggregation and disaggregation of fibrillogenesis with IC50 values of 3 92 and 7 19 M respectively A aggregation-IN-1 also inhibits malondialdehyde formation augments the intracellular reduced glutathione (GSH) levals and inhibits caspase 3 in neuronal cells[1]
Catalog No. 50-002-06021 Supplier Medchemexpress LLC Supplier No. 250MGHYW117986
A aggregation-IN-1 (Compound 1b) is an amyloid-beta precursor protein inhibitor A aggregation-IN-1 inhibits amyloid-beta aggregation and disaggregation of fibrillogenesis with IC50 values of 3 92 and 7 19 M respectively A aggregation-IN-1 also inhibits malondialdehyde formation augments the intracellular reduced glutathione (GSH) levals and inhibits caspase 3 in neuronal cells[1]