Learn More
Bachem Phalloidin Trifluoroacetate
SDP
Supplier:  Bachem 4095642.0001
Encompass_Preferred
Phalloidin Trifluoroacetate, Cyclo(-Ala-D-Thr-Cys-cis-Hyp-Ala-Trp-(4R)-4,5-dihydroxy-Leu), 17466-45-4, C35H48N8O11S, Mr 788.87, 1mg. Bicyclic(Ala-DThr-Cys-cis-4-hydroxy-Pro-Ala-2-mercapto-Trp-4,5-dihydroxy-Leu)(S-3→6), NSC 523214. A bicyclic 7 amino acid peptide toxin originally isolated from the poisonous mushroom Amanita phalloides. Phalloidin binds specifically to the interface between F-actin subunits.
Catalog No. 50-194-7504