Learn More
Cayman Chemical (±)-α-Bisabolol, Purity: ≥95% (mixture of isomers), CAS: 515-69-5, 50 g
SDP
Catalog No. 502990759
Encompass_Preferred
A sesquiterpene alcohol with anticancer activity; induces apoptosis in, and is cytotoxic to, T67 and U87 glioma cells from 3.5-10 µM; cytotoxic to primary ALL and AML cells (IC50s = 14-65 µM)
Catalog No. 50-299-0759 Supplier Cayman Chemical Supplier No. 3139750 G
A sesquiterpene alcohol with anticancer activity; induces apoptosis in, and is cytotoxic to, T67 and U87 glioma cells from 3.5-10 µM; cytotoxic to primary ALL and AML cells (IC50s = 14-65 µM)