Learn More
Selleck Chemical LLC a-bisabolol
SDP
Catalog No. 5000000789
Encompass_Preferred
-bisabolol is a small plant-derived oily sesquiterpene alcohol with some anti-inflammatory and even anti-microbial properties
Catalog No. 50-000-00789 Supplier Selleck Chemical LLC Supplier No. S613625UL
-bisabolol is a small plant-derived oily sesquiterpene alcohol with some anti-inflammatory and even anti-microbial properties