Learn More
Bachem H-Gly-Leu-OH
SDP
Supplier:  Bachem 4000534.0100
Encompass_Preferred
H-Gly-Leu-OH, 869-19-2, C8H16N2O3, Mr 188.23, 100g. A bitter-tasting dipeptide, which activates the human bitter receptors hTAS2Rs. GL is a good substrate for dipeptide hydrolase activities in rat intestinal mucosa and human kidney.
Catalog No. 50-194-6830