Learn More
Bachem Fmoc-D-Asp(OtBu)-(Hmb)Gly-OH
SDP
Supplier:  Bachem 4035340.0001
Encompass_Preferred
Fmoc-D-Asp(OtBu)-(Hmb)Gly-OH, 1926163-00-9, C33H36N2O9, Mr 604.67, 1g. Fmoc-D-Asp(OtBu)-N-(2-hydroxy-4-methoxy-Bzl)-Gly-OH. A building block for the synthesis of peptides containing the D-Asp-Gly motif avoiding base-catalyzed aspartimide formation.
Catalog No. 50-194-7166