Learn More
Bachem L-threo-Phenylserine
SDP
Supplier:  Bachem 4064845.0005
Encompass_Preferred
L-threo-Phenylserine, 6254-48-4, C9H11NO3, Mr 181.19, 5g. (2S,3R)-2-Amino-3-hydroxy-3-phenylpropionic acid. A building block of peptide antibiotics and a valuable chiral synthon. Crich and Banerjee used it for obtaining b-mercaptophenylalanine. Introduction of this Phe derivative allowed native chemical ligation at Phe followed by desulfuration.
Catalog No. 50-194-7265