Learn More
American Radiolabeled Chemicals Inc A-BUNGAROTOXIN 125I 250UC
SDP
Catalog No. 50901028
Encompass
a-Bungarotoxin, Tyr54-125I 250uCi in buffered solution (pH 7.2), 0.25 mCi/ml, S.A. 2200 Ci/mmol, M.W. 7984
Catalog No. 50-901-028 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 0146250UC
a-Bungarotoxin, Tyr54-125I 250uCi in buffered solution (pH 7.2), 0.25 mCi/ml, S.A. 2200 Ci/mmol, M.W. 7984