Learn More
American Radiolabeled Chemicals Inc A-BUNGAROTOXIN 125I 50UC
SDP
Catalog No. 50901029
Encompass
a-Bungarotoxin, Tyr54-125I 50uCi in buffered solution (pH 7.2), 0.25 mCi/ml, S.A. 2200 Ci/mmol, M.W. 7984
Catalog No. 50-901-029 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 014650UC
a-Bungarotoxin, Tyr54-125I 50uCi in buffered solution (pH 7.2), 0.25 mCi/ml, S.A. 2200 Ci/mmol, M.W. 7984