Learn More
Biotium Alpha-Bungarotoxin CF488A
Catalog No. NC1493907
Encompass_Preferred
Fluorescent conjuates of -Bungarotoxin bind to the nicotinic acetylcholine receptor with high affinity, and thus can be used to image the distribution of the receptor at the neuromuscular junctions. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye (Ex/Em 490/515 nm) is a superior alternative to FITC (fluorescein) due to its exceptional brightness and photostability.
Catalog No. NC1493907 Supplier Biotium Supplier No. 00005100UG
Fluorescent conjuates of -Bungarotoxin bind to the nicotinic acetylcholine receptor with high affinity, and thus can be used to image the distribution of the receptor at the neuromuscular junctions. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye (Ex/Em 490/515 nm) is a superior alternative to FITC (fluorescein) due to its exceptional brightness and photostability.