Learn More
Grainger Starrett A/C Adaptor, 110 V, 2700 Series, 1 ea
Catalog No. 502787660
Encompass_Preferred
The A/C adaptor is designed to provide a reliable power source for 2700 Series electronic indicators. This unit ensures continuous operation by serving as a dedicated power supply in a 110 V configuration.
  • Designed for use with 2700 series electronic indicators
  • Operates at 110 V
  • Facilitates benchtop or stationary use when battery power is not required
Catalog No. 50-278-7660 Supplier Grainger Supplier No. 197R96
The A/C adaptor is designed to provide a reliable power source for 2700 Series electronic indicators. This unit ensures continuous operation by serving as a dedicated power supply in a 110 V configuration.
  • Designed for use with 2700 series electronic indicators
  • Operates at 110 V
  • Facilitates benchtop or stationary use when battery power is not required