Learn More
Bioworld ACE Acetate Buffer, 50mM, pH 4.5, 100 mL
Catalog No. 50198610
Encompass_Preferred
Product DescriptionA.C.E.™ acetate buffer has a concentration of 50 mM with a pH level of 4.5, and is formulated for capillary electrophoresis. Buffer is ideal for the separation of various biomolecules including proteins. A.C.E.™ Buffers deliver consistent and reproducible results because they are manufactured with the highest quality reagents in nuclease free water.
Catalog No. 50-198-610 Supplier Bioworld Supplier No. 214200031
Product DescriptionA.C.E.™ acetate buffer has a concentration of 50 mM with a pH level of 4.5, and is formulated for capillary electrophoresis. Buffer is ideal for the separation of various biomolecules including proteins. A.C.E.™ Buffers deliver consistent and reproducible results because they are manufactured with the highest quality reagents in nuclease free water.