Learn More
Bioworld A.C.E. Buffer TINTED (10X), 50 mL
Supplier:  Bioworld 214200041
Encompass_Preferred
10X running buffer concentrate.Tinted light pink for easy differentiation from other solvents in the sequencing apparatus.
Catalog No. 50-198-612