Learn More
Bioworld A.C.E. Buffer TINTED (1X), 1 L
Supplier:  Bioworld 214200052
Encompass_Preferred
Ready-to-use 1X running buffer.Tinted light pink for easy differentiation from other solvents in the sequencing apparatus.
Catalog No. 50-198-615