Learn More
Bioworld ACE MES Buffer, 50mM, pH 6.0, 100 mL
Catalog No. 50198616
Encompass_Preferred
Product DescriptionA.C.E.™ MES (compound 2-(N-morpholino)ethanesulfonic acid) buffer has a concentration of 50 mM with a pH level of 6.0. The buffer is biotechnology grade and is formulated for capillary electrophoresis. Buffer is made with 3D water and filtered through 0.22 µ filter. Product is shipped at room temperature.
Catalog No. 50-198-616 Supplier Bioworld Supplier No. 214200061
Product DescriptionA.C.E.™ MES (compound 2-(N-morpholino)ethanesulfonic acid) buffer has a concentration of 50 mM with a pH level of 6.0. The buffer is biotechnology grade and is formulated for capillary electrophoresis. Buffer is made with 3D water and filtered through 0.22 µ filter. Product is shipped at room temperature.