Learn More
Bioworld A.C.E. Phosphate Buffer, 50 mM, (pH 9.3), 100 mL
Catalog No. 50198633
Encompass_Preferred
These convenient 50 mM premixed and ready-to-use A.C.E.™ Phosphate Buffers have been specially formulated for Capillary Electrophoresis. They are ideal for the separation of various biomolecules including proteins. Manufactured with the highest quality reagents in nucleasefree water, AMRESCO's A.C.E.™ Buffers deliver consistent and reproducible results.
Catalog No. 50-198-633 Supplier Bioworld Supplier No. 214200141
These convenient 50 mM premixed and ready-to-use A.C.E.™ Phosphate Buffers have been specially formulated for Capillary Electrophoresis. They are ideal for the separation of various biomolecules including proteins. Manufactured with the highest quality reagents in nucleasefree water, AMRESCO's A.C.E.™ Buffers deliver consistent and reproducible results.