Learn More
Bioworld ACE TBE w/ 7M Urea, pH 8.6, 250 mL
Catalog No. 50198636
Encompass_Preferred
Product DescriptionA.C.E. TM TBE (Tris-Borate-EDTA) buffer comes with 7M of Urea with a pH level of 8.6. A.C.E. TM buffers deliver consistent and reproducible results because they are manufactured with the highest quality reagents in nuclease free water. Product is shipped at room temperature.
Catalog No. 50-198-636 Supplier Bioworld Supplier No. 214200152
Add to Cart
Add to Cart
Product DescriptionA.C.E. TM TBE (Tris-Borate-EDTA) buffer comes with 7M of Urea with a pH level of 8.6. A.C.E. TM buffers deliver consistent and reproducible results because they are manufactured with the highest quality reagents in nuclease free water. Product is shipped at room temperature.