Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ A.C.T.™ I Culture Transport

Catalog No. R124100 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R124100 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R124100 Supplier Thermo Scientific™ Supplier No. R124100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.

Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples (swab not included).

  • With semi-solid, nonnutritive medium for use with swab or fluid samples
  • Fluids may be introduced through septum in cap
TRUSTED_SUSTAINABILITY

Specifications

Sterility Sterile
Description Single tube
For Use With (Application) Healthcare
Quantity 10/Pk.
Storage Requirements Store in original container at 15°C to 30°C until used.
Tube Size 15 x 105 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.