Learn More
Thermo Scientific™ A.C.T.™ I Culture Transport
Catalog No. R12401 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
R12401 20/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R12401 Supplier Thermo Scientific™ Supplier No. R12401
This item is not returnable. View return policy
Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
  • With semi-solid, nonnutritive medium for use with swab or fluid samples
  • Fluids may be introduced through septum in cap
TRUSTED_SUSTAINABILITY
Specifications
Description A.C.T. I
For Use With (Application) Healthcare
Quantity 20/Pk.
Tube Size 16 x 125 mm