Learn More
Bachem a-CGRP (8-37) (mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4034544.0005
Encompass_Preferred
a-CGRP (8-37) (mouse, rat) Trifluoroacetate, H-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asp-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH2, 129121-73-9, C138H224N42O41, Mr 3127.56, 5mg. Calcitonin Gene-Related Peptide I (8-37) (mouse, rat), CGRP-I (8-37) (mouse, rat). a-CGRP (8-37), a competitive CGRP1 receptor antagonist, exhibited hypothermic and antinociceptive effects. CGRP-I (8-37) antagonized the effects of amylin and amylin (1-8) on osteoblasts but was substantially less potent in this regard than amylin (8-37). Furthermore, it inhibited the evoked discharge frequency of wide dynamic range neurons in dorsal horn of the spinal cord in rats.
Catalog No. 50-259-898