Learn More
Ibidi USA -Slide 8 Well high
SDP
Catalog No. NC1930679
Encompass
A chambered coverslip with 8 individual wells, high walls. All-in-one 8 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Hish-resolution imaging through the #1.5 polymer coverslip bottom with the highest optical quality. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC1930679 Supplier Ibidi USA Supplier No. 80806
A chambered coverslip with 8 individual wells, high walls. All-in-one 8 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Hish-resolution imaging through the #1.5 polymer coverslip bottom with the highest optical quality. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)