Learn More
Dravon Medical Inc A-Clamp, Non-Sterile, Plastic Disposable, Blue, 25/pk
SDP
Catalog No. NC9728735
Encompass
(True Blue) A-Clamp,25 per pack,Material: Special formulation of glass-filled nylon with metal rivet,The A-clamp can be autoclaved up to 132degree C,gas sterilized,gamma radiated,or electron beam sterilized and will withstand most cold ster
Catalog No. NC9728735 Supplier Dravon Medical Inc Supplier No. A110/25
(True Blue) A-Clamp,25 per pack,Material: Special formulation of glass-filled nylon with metal rivet,The A-clamp can be autoclaved up to 132degree C,gas sterilized,gamma radiated,or electron beam sterilized and will withstand most cold ster