Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A Closer Look at Aspirin Lab
SCI_ED

Catalog No. S67403
Change view
Click to view available options
For a closer look at Asprin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67403 For a closer look at Asprin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67403 Supplier Neo/Sci Corporation Supplier No. 201773

Study the world's most common over-the-counter medication.

Learn what this pain-reliever and blood thinner is made of, what it does and how brands differ.

Teaches students about:

  • Calculating the percent of binder in tablets
  • Determining binder composition
  • Investigating tablet dissolution
  • Determining pH of tablet solution
  • Testing for salicylates
  • Determining cost-benefit relationships among attributes

Order Info

Class size: 40 students

Specifications

For Use With (Application) To study binder composition and percent, tablet dissolution, ph and testing for salicylates
Class Size 40 Students
Product Type For A Closer Look At Asprin Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.